Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUV39H2 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001140 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001140 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001140 Supplier Novus Biologicals Supplier No. NB1001140
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 2 publications

SUV39H2 Polyclonal specifically detects SUV39H2 in Human, Rat samples. It is validated for Chemotaxis, ChIP Assay, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Peptide ELISA, Western Blot.

Specifications

Antigen SUV39H2
Applications Chemotaxis, ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Peptide ELISA detection limit 1:8000, Chromatin Immunoprecipitation (ChIP), Immunocytochemistry 1:100
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_078946.1
Gene Alias FLJ23414Histone H3-K9 methyltransferase 2, H3-K9-HMTase 2, histone-lysine N-methyltransferase SUV39H2, KMT1BEC 2.1.1.43, Lysine N-methyltransferase 1B, Su(var)3-9 homolog 2, Suppressor of variegation 3-9 homolog 2, suppressor of variegation 3-9 homolog 2 (Drosophila)
Gene Symbols SUV39H2
Host Species Goat
Immunogen Peptide with sequence CKCGAVTCRGYLN corresponding to C-Terminus according to NP_078946.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 79723
Test Specificity Tested in rat. Expected from sequence similarity: Human, Mouse, Rat, Dog, Pig, Cow.
Target Species Human, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.