Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUV420h1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP197313SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP197313SS 0.025 mL
NBP197313 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP197313SS Supplier Novus Biologicals Supplier No. NBP197313SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SUV420h1 Polyclonal specifically detects SUV420h1 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Western Blot (Negative).

Specifications

Antigen SUV420h1
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.16 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 2-5 ug/ml, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:200, Western Blot (Negative)
Formulation PBS and 30% Glycerol with 0.05% Sodium Azide
Gene Accession No. Q4FZB7, Q3U8K7
Gene Alias C630029K18Rik, CGI85, CGI-85, EC 2.1.1.43, histone-lysine N-methyltransferase SUV420H1, KMT5B, KMT5B Lysine N-methyltransferase 5B, MGC118906, MGC118909, MGC21161, MGC703, Su(var)4-20 homolog 1, Suppressor of variegation 4-20 homolog 1, suppressor of variegation 4-20 homolog 1 (Drosophila), suv4-20h1
Gene Symbols KMT5B
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human SUV420h1 protein (between residues 450-500) [UniProt Q4FZB7]
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, Epigenetics, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 51111
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.