Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Suz12 Monoclonal Antibody (2AO9), Active Motif™
SDP

Mouse Monoclonal Antibody

$135.30 - $405.90

Specifications

Antigen Suz12
Clone 2AO9
Concentration 1 mg/mL
Applications ChIP Assay
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-2923
SDP
Encompass
View Documents
Active Motif
39878
10 μg
Each for $135.30
Only null left
Add to Cart
 
50-199-2922
SDP
Encompass
View Documents
Active Motif
39877
100 μg
Each for $405.90
Only null left
Add to Cart
 
Description

Description

SUZ12 (polycomb protein SUZ12) is a zinc finger gene and has been identified at the breakpoints of a recurrent chromosomal translocation reported in endometrial stromal sarcoma. Recombination of these breakpoints results in the fusion of this gene and JAZF1. SUZ12 is also a component of the PRC2/EED-EZH2 complex which methylates Lys-9 (H3K9me) and Lys-27 (H3K27me) of histone H3 leading to transcriptional repression of the affected target gene. The protein encoded by this gene contains a zinc finger domain in the C terminus of the coding region.
Specifications

Specifications

Suz12
1 mg/mL
Monoclonal
Liquid
RUO
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
2610028O16Rik; AI195385; AU016842; AW536442; ChET 9 protein; CHET9; Chromatin precipitated E2F target 9 protein; D11Ertd530e; JJAZ1; Joined to JAZF1 protein; KIAA0160; mKIAA0160; Polycomb protein Suz12; suppressor of zeste 12 homolog (Drosophila); Suppressor of zeste 12 protein homolog; SUZ12; SUZ12 polycomb repressive complex 2 subunit
SUZ12
IgG2b
Protein G
Antibody
2AO9
ChIP Assay
Unconjugated
Mouse
Human
Q15022
23512
This Suz12 antibody was raised against a recombinant protein corresponding to full-length human Suz12.
Primary
-20°C, Avoid Freeze/Thaw Cycles
SUZ12
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.