Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUZ12 Rabbit anti-Human, Mouse, Rat, Clone: 1M3V2, Novus Biologicals™
SDP

Catalog No. NB167858 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167858 20 μg
NB167857 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB167858 Supplier Novus Biologicals Supplier No. NBP31638320UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

SUZ12 Monoclonal antibody specifically detects SUZ12 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen SUZ12
Applications Western Blot
Classification Monoclonal
Clone 1M3V2
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias ChET 9 protein, CHET9polycomb protein SUZ12, Chromatin precipitated E2F target 9 protein, JJAZ1Joined to JAZF1 protein, KIAA0160Suppressor of zeste 12 protein homolog, suppressor of zeste 12 homolog (Drosophila)
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-96 of human SUZ12 (Q15022).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Chromatin Modifiers
Primary or Secondary Primary
Gene ID (Entrez) 23512
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.