Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SV2A Antibody, Novus Biologicals™
SDP

Catalog No. NB10041402 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10041402 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10041402 Supplier Novus Biologicals Supplier No. NB10041402
Add to Cart
Add to Cart

Goat Polyclonal Antibody

SV2A Polyclonal specifically detects SV2A in Human, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Peptide ELISA.

Specifications

Antigen SV2A
Applications Immunocytochemistry, Immunofluorescence, Peptide ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 10 ug/mL, Peptide ELISA Detection limit 1:1000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_055664.2
Gene Alias KIAA0736SV2, synaptic vesicle glycoprotein 2A
Gene Symbols SV2A
Host Species Goat
Immunogen Peptide with sequence C-TVELYPSDKRTTA corresponding to internal region according to NP_055664.3.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9900
Test Specificity Cross reacts with mammal. Expected from sequence similarity: Human, Mouse, Rat, Pig, Cow and Dog.
Target Species Human, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.