Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SV40 Large T Antigen Ab-1 Mouse Monoclonal Antibody, Epredia™

Epredia™ SV40 Large T Antigen Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Specifications

Antigen SV40 Large T Antigen Ab-1
Clone PAb101; same as clone 412
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Concentration Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Concentration Price Quantity & Availability  
MS1832P0
View Documents
Epredia
MS1832P0
100 μL 200μg/mL N/A N/A N/A
MS1832P1
View Documents
Epredia
MS1832P1
500 μL 200μg/mL N/A N/A N/A
MS1832P
View Documents
Epredia
MS1832P
1 mL 200μg/mL N/A N/A N/A
MS1832P1ABX
View Documents
Epredia
MS1832P1ABX
100 μL 1mg/mL N/A N/A N/A
MS1832PABX
View Documents
Epredia
MS1832PABX
200 μL 1mg/mL N/A N/A N/A
Description

Description

The simian virus 40 large T antigen plays important role in SV40 transfection and transformation of the cells. It binds to Rb and p53 suppressor proteins leading to the inactivation of suppressor pathway.

Host Species: Mouse

Clone: PAb101; originally clone 412

Isotype: IgG2a1

Species Reactivity: SV40 virus.

Epitope: C-terminal

Immunogen: SV40-transformed mouse line B4

Molecular Weight: 94kDa

Positive Control: SV40-transfected cells

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

SV40 Large T Antigen Ab-1
Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
Mouse
Infectious Disease
SV40 transformed mouse line B4
PAb101; same as clone 412
Monoclonal
Concentrated
RUO
Simian virus 40 (SV40)
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.