Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SV40 Large T Antigen Antibody (LP7N), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB309599 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309599 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309599 Supplier Novus Biologicals Supplier No. NBP250312MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SV40 Large T Antigen Monoclonal antibody specifically detects SV40 Large T Antigen in Virus samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen SV40 Large T Antigen
Applications Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone LP7N
Conjugate mFluor Violet 610 SE
Dilution Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Large T antigen, Small T antigen
Host Species Mouse
Immunogen Detergent insoluble fraction of SVK14 cell line (SV40 transformed neonatal keratinocytes)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.