Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SV40 Small T antigen Antibody (pab 280) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261248
Description
SV40 Small T antigen Monoclonal specifically detects SV40 Small T antigen in Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
SV40 Small T antigen | |
Monoclonal | |
0.9 mg/ml | |
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
Mouse | |
Protein A purified | |
Cancer, Virology Bacteria and Parasites | |
Virus | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
pab 280 | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
A purified Pab 419-SV40 Small T antigen | |
0.1 mL | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction