Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SV40 Small T antigen Antibody (pab 280) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10 - $426.00
Specifications
Antigen | SV40 Small T antigen |
---|---|
Clone | pab 280 |
Concentration | 0.9 mg/ml |
Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Description
SV40 Small T antigen Monoclonal specifically detects SV40 Small T antigen in Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
SV40 Small T antigen | |
0.9 mg/ml | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Virus | |
A purified Pab 419-SV40 Small T antigen | |
Primary |
pab 280 | |
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
Monoclonal | |
Purified | |
Cancer, Virology Bacteria and Parasites | |
PBS with 0.02% Sodium Azide | |
IgG1 | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title