Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SV40 T-antigen Antibody (Pab 430), PE, Novus Biologicals™
SDP

Catalog No. NB309238 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309238 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309238 Supplier Novus Biologicals Supplier No. NBP250598PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SV40 T-antigen Monoclonal antibody specifically detects SV40 T-antigen in Virus samples. It is validated for Western Blot, Immunoprecipitation, Radioimmune Assays

Specifications

Antigen SV40 T-antigen
Applications Western Blot, Immunoprecipitation, Radioimmune Assays (RIA)
Classification Monoclonal
Clone Pab 430
Conjugate PE
Dilution Western Blot, Immunoprecipitation, Radioimmunoassay
Formulation PBS
Host Species Mouse
Immunogen SV40 transformed mouse cells
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.