Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ SW 033291

missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
Description
SW 033291 is a high affinity 15-hydroxyprostaglandin dehydrogenase (15-PGDH) inhibitor (Ki = 0.1 nM). Increases PGE2 levels in A549 cells in vitro, as well as in bone marrow, colon, lung and liver in mice. Promotes hematopoiesis, recovery from DSS-induced colitis, and hepatocyte proliferation after partial hepatectomy in mice.
Specifications
Specifications
| Chemical Name or Material | 2-(Butylsulfinyl)-4-phenyl-6-(2-thienyl)-thieno[2,3-b]pyridin-3-amine |
| CAS | 459147-39-8 |
| Quantity | 10 mg |
| Target | Dehydrogenase Inhibitors |
| Molecular Formula | C21H20N2OS3 |
| Formula Weight | 412.59 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction