Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SWAP-70, Rabbit anti-Human, Polyclonal, Millipore Sigma™
Rabbit Polyclonal Antibody
$192.34 - $482.80
Specifications
Antigen | SWAP-70 |
---|---|
Dilution | Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution from a representative lot detected SWAP-70 in human tonsil and human lung tissue sections. |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABC168825UG
|
MilliporeSigma
ABC168825UG |
25 μg |
Each for $192.34
|
|
|||||
ABC1688100
|
MilliporeSigma
ABC1688100UG |
100 μg |
Each for $482.80
|
|
|||||
Description
Anti-SWAP-70, Cat. No. ABC1688, is a rabbit polyclonal antibody that detects Switch-associated protein 70 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Switch-associated protein 70 (UniProt: Q9UH65; also known as SWAP-70) is encoded by the SWAP70 (also known as KIAA0640, HSPC321) gene (Gene ID: 23075) in human. SWAP-70 is a phosphatidylinositol 3,4,5-trisphosphate (PIP3)-dependent guanine nucleotide exchange factor (GEF) that, independently of RAS, transduces signals from tyrosine kinase receptors to RAC. SWAP-70 regulates the actin cytoskeleton as an effector or adapter protein in response to agonist stimulated phosphatidylinositol (3,4)-bisphosphate production and cell protrusion. It is expressed only in mature B-cells including those associated with mucosa-associated tissue and bronchus-associated tissue. In resting B-cells it is localized mainly in the cytoplasm and upon cell activation it is recruited to the plasma membrane and then translocates to the nucleus. In activated, class-switching B-cells it is associated with membrane IgG, but not IgM. It is also widely expressed in spleen, kidney, lung, and liver. SWAP-70 has an N-terminal putative EF-hand domain, a pleckstrin homology (PH) domain (aa 210-306) that binds phosphatidylinositol 3,4,5-trisphosphate and is responsible for membrane localization, a tri-partite coiled-coil region (aa 316-539), and a C-terminal F-actin-binding site through which it specifically binds non-muscle actin. (Ref.: Chacón-Martínez, CA et al. (2103). J. Biol. Chem. 288(40); 28687-28703).Specifications
SWAP-70 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
RUO | |
Human | |
Switch-associated protein 70 | |
SWAP70;KIAA0640;HSPC321 | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. | |
∽70 kDa observed; 69.00 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution from a representative lot detected SWAP-70 in human tonsil and human lung tissue sections. | |
Polyclonal | |
Rabbit | |
Apoptosis & Cancer | |
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
NP_001284643 | |
KLH-conjugated linear peptide corresponding to 12 amino acids from the N-terminal region of human Switch-associated protein 70. | |
Affinity Purified | |
This rabbit polyclonal antibody detects human Switch-associated protein 70. It targets an epitope with in 12 amino acids from the N-terminal region. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title