Learn More
Thermo Scientific™ SWAP-IT™ GC-MS Interface System Accessories
Catalog No. 290SG001
Click to view available options
Type:
Ferrule
SWAP-Tight Ferrule 0.1 to 0.25 mm
SWAP-Tight Ferrule 0.32 mm
SWAP-Tight Ferrule, Blank
SWAP-Tight Nut
Transfer Line Nut
Find the right nut and ferrule to use with the powerful Thermo Scientific™ SWAP-IT™ GC-MS Interface system.
  • Specially deactivated , robust stainless steel transfer capillary provides an interface between the analytical column and the MS detector
  • Small ID of transfer capillary ensures exposure to air and water is reduced to a minimum during disconnection of the analytical column
  • The system works without the requirement for additional He-purge/make-up gas flow
  • Finger-tight connections make exchanging the analytical column quick and easy without the need for tools
  • Includes finger-tight connections for S/SL injector to provide complete finger-tight solution
TRUSTED_SUSTAINABILITY
Specifications
Type Transfer Line Nut
Quantity 1/Ea.
For Use With (Equipment) SWAP-IT GC-MS Interface System for DSQ GC-MS, DSQ II GC-MS, PolarisQ GC-MS, ITQ GC-MS Only
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.