Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SWAP-IT™ GC-MS Interface System Accessories

Catalog No. 290SG032
Click to view available options
Type:
Ferrule
SWAP-Tight Ferrule 0.1 to 0.25 mm
SWAP-Tight Ferrule 0.32 mm
SWAP-Tight Ferrule, Blank
SWAP-Tight Nut
Transfer Line Nut

Find the right nut and ferrule to use with the powerful Thermo Scientific™ SWAP-IT™ GC-MS Interface system.

  • Specially deactivated , robust stainless steel transfer capillary provides an interface between the analytical column and the MS detector
  • Small ID of transfer capillary ensures exposure to air and water is reduced to a minimum during disconnection of the analytical column
  • The system works without the requirement for additional He-purge/make-up gas flow
  • Finger-tight connections make exchanging the analytical column quick and easy without the need for tools
  • Includes finger-tight connections for S/SL injector to provide complete finger-tight solution
TRUSTED_SUSTAINABILITY

Specifications

Type SWAP-Tight Ferrule 0.32 mm
Quantity 10 Pk.
For Use With (Equipment) SWAP-IT GC-MS Interface System

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.