Learn More
Thermo Scientific™ SWAP-IT™ GC-MS Interface System Accessories
Catalog No. p-6062425
Change view
Click to view available options
Type:
Ferrule
SWAP-Tight Ferrule 0.1 to 0.25 mm
SWAP-Tight Ferrule 0.32 mm
SWAP-Tight Ferrule, Blank
SWAP-Tight Nut
Transfer Line Nut
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
29-0SG-000 SWAP-Tight Ferrule, Blank
29-0SG-001 Transfer Line Nut
29-0SG-002 SWAP-Tight Nut
29-0SG-021 Ferrule
29-0SG-025 SWAP-Tight Ferrule 0.1 to 0.25 mm
29-0SG-032 SWAP-Tight Ferrule 0.32 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 29-0SG-000 Supplier Thermo Scientific™ Supplier No. 290SG000
Only null left
Add to Cart
Add to Cart
Find the right nut and ferrule to use with the powerful Thermo Scientific™ SWAP-IT™ GC-MS Interface system.
  • Specially deactivated , robust stainless steel transfer capillary provides an interface between the analytical column and the MS detector
  • Small ID of transfer capillary ensures exposure to air and water is reduced to a minimum during disconnection of the analytical column
  • The system works without the requirement for additional He-purge/make-up gas flow
  • Finger-tight connections make exchanging the analytical column quick and easy without the need for tools
  • Includes finger-tight connections for S/SL injector to provide complete finger-tight solution
TRUSTED_SUSTAINABILITY
Specifications
Type SWAP-Tight Ferrule, Blank
Quantity 10 Pk.
For Use With (Equipment) SWAP-IT GC-MS Interface System
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.