Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SWAP70 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056430 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056430 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056430 Supplier Novus Biologicals Supplier No. NB10056430
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SWAP70 Polyclonal specifically detects SWAP70 in Human samples. It is validated for Western Blot.

Specifications

Antigen SWAP70
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml
Formulation PBS containing 0.2% gelatin with 0.05% Sodium Azide
Gene Accession No. Q9UH65
Gene Alias KIAA0640SWAP-70FLJ39540, SWAP switching B-cell complex 70kDa subunit, switch-associated protein 70
Gene Symbols SWAP70
Host Species Rabbit
Immunogen A portion of amino acids 300-350 of human SWAP-70 was used as the immunogen.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23075
Test Specificity Reacts with Human. Predicted to react with Bovine, Chimpanzee, Dog and Mouse.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.