Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Swi6 Antibody, Novus Biologicals™
SDP

Catalog No. NB600932 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600932 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600932 Supplier Novus Biologicals Supplier No. NB600932
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Swi6 Polyclonal specifically detects Swi6 in S. pombe samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Swi6
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, ELISA 1:5000-1:30000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. P40381
Gene Alias Chromatin-associated swi6 protein; Regulatory protein SWI6
Gene Symbols SWI6
Host Species Rabbit
Immunogen This Swi6 Antibody was prepared from whole rabbit antiserum produced by repeated immunizations with a synthetic peptide corresponding aa 314-328 of S.pombe Swi6 protein. (Uniprot: P40381)
Molecular Weight of Antigen 37.29 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 850879
Test Specificity Swi6
Target Species S. pombe
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.