Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Swine Influenza A H1N1 Hemagglutinin Antibody (3E9H5), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB199771 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB199771 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB199771 Supplier Novus Biologicals Supplier No. NBP241329AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Swine Influenza A H1N1 Hemagglutinin Monoclonal antibody specifically detects Swine Influenza A H1N1 Hemagglutinin in Virus samples. It is validated for ELISA

Specifications

Antigen Swine Influenza A H1N1 Hemagglutinin
Applications ELISA
Classification Monoclonal
Clone 3E9H5
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Antibody was raised against a synthetic peptide containing the sequence specific to the novel S-OIV A H1N1 strain hemagglutinin protein. Amino Acid Squence: STSADQQSLYQNAD
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.