Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Swine Influenza A H1N1 NS1 Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB198761 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB198761 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB198761 Supplier Novus Biologicals Supplier No. NBP241342JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Swine Influenza A H1N1 NS1 Polyclonal antibody specifically detects Swine Influenza A H1N1 NS1 in Virus samples. It is validated for ELISA

Specifications

Antigen Swine Influenza A H1N1 NS1
Applications ELISA
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide from the swine-Origin Influenza NS1. The immunogen is located within the last 50 amino acids of Swine H1N1 Nonstructural Protein 1. Amino Acid Squence: DENGRPSLPPE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.