Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYBR GreenER™ qPCR SuperMix for ABI PRISM™ Instrument

Incorporate the latest high-performance technology to provide the most reliable gene expression data from your ABI 7300, 7000, 7700 or 7900HT instrument.

$391.00 - $1664.00

Specifications

Concentration 2X
Detection Method SYBR
GC-Rich PCR Performance High
PCR Method qPCR
For Use With (Equipment) 7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System
View More Specs

Products 2
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
11-760-500
Encompass_Preferred
View Documents
Invitrogen™
11760500
500 Reactions
Each for $1,664.00
Only null left
Add to Cart
 
11-760-100
Encompass_Preferred
View Documents
Invitrogen™
11760100
100 Reactions
Each for $391.00
Only null left
Add to Cart
 
Description

Description

The SYBR™ GreenER™ qPCR SuperMix kits include a dsDNA-binding dye that produces a brighter signal and significantly less PCR inhibition, for improved qPCR performance over a broad dynamic range.

Consistent Specificity and Low-Copy Detection: The SYBR™ GreenER™ qPCR reagent system minimizes the formation of nonspecific products, including primer-dimers, for greater accuracy across a wide range of targets. The reliability of the SYBR™ GreenER™ qPCR reagent system also translates to greater sensitivity. With SYBR™ GreenER™ qPCR SuperMix, you can consistently detect fewer than 10 copies of a target in genomic DNA.

  • The SYBR™ GreenER™ system is specially formulated to offer the best sensitivity, specificity, and reproducibility
  • A novel dsDNA-binding dye - exhibits a brighter signal for increased sensitivity and less PCR inhibition than original SYBR™ Green I, while using the same instrument filters and settings
  • An optimized buffer system - improves sensitivity and provides excellent long-term stability
  • Uracil DNA glycosylase - reduces carryover contamination in qPCR

Order Info

Shipping Conditions: Dry Ice

Specifications

Specifications

2X
High
7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System
Real Time PCR SYBR Master Mix
DNA (Genomic), cDNA
SYBR
qPCR
Platinum™, SYBR™ GreenER™
Gene Expression
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.