Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SYDE1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15760720 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15760720 20 μL
NBP157607 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15760720 Supplier Novus Biologicals Supplier No. NBP15760720UL

Rabbit Polyclonal Antibody

SYDE1 Polyclonal specifically detects SYDE1 in Human samples. It is validated for Western Blot, Knockout Validated.

Specifications

Antigen SYDE1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS & 2% Sucrose. with No Preservative
Gene Accession No. Q6ZW31
Gene Alias EC 2.4.1.25, EC 2.6.1.9, FLJ13511, Protein syd-1 homolog 1, rho GTPase-activating protein SYDE1, synapse defective 1, Rho GTPase, homolog 1 (C. elegans), Synapse defective protein 1 homolog 1,7h3
Gene Symbols SYDE1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to SYDE1(synapse defective 1, Rho GTPase, homolog 1 (C. elegans)) The peptide sequence was selected from the C terminal of SYDE1. Peptide sequence PYLRPKRQPPLHLPLADPEVVTRPRGRGGPESPPSNRYAGDWSVCGRDFL.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85360
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.