Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Syk Monoclonal Antibody (4D10.1), APC, eBioscience™
GREENER_CHOICE

Catalog No. 5015288
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-152-88 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-152-88 Supplier Invitrogen™ Supplier No. 17669642
  • $418.00
    $313.65 / Each of 1

    Save $104.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: This 4D10.1 monoclonal antibody recognizes human Syk, a 72-kDa member of the Syk/ZAP-70 family of non-receptor protein tyrosine kinases. Syk is expressed most highly in B cells, with lower expression in immature thymocytes, mast cells, and platelets. Syk can also be detected in fibroblasts, epithelial cells, hepatocytes, endothelial cells, and neuronal cells. This protein is a major component of signaling cascades downstream of the B and T cell antigen receptors and plays an essential role in lymphocyte development. Upon recruitment to immunoreceptor tyrosine-based activation motifs (ITAMs) on the antigen receptors, Syk is activated by phosphorylation on multiple tyrosines. Once activated, Syk phosphorylates proteins such as phospholipase C gamma and BLNK/SLP-65. Finally, abnormal Syk expression has been linked to tumor cell migration and invasion in several cancers. Applications Reported: This 4D10.1 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This 4D10.1 antibody has been pre-titrated and tested by intracellular staining using the Foxp3 buffer system and flow cytometric analysis of normal peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 n...

Spleen tyrosine kinase (SYK), along with ZAP70, are cytoplasmic tyrosine kinases that are viewed as potential druge targets for allergic disorders and autoimmune diseases. SYK may also play a role in cancer. Syk translocates to the plasma membrane upon B cell antigen receptor (BCR) or the high-affinity IgE receptor (FcepsilonRI) triggering, and phosphorylates downstream adaptor proteins, thereby providing docking sites for initiation of subsequent signaling pathways, such as calcium mobilization, cytoskeleton remodeling, or transcription of specific genes. Syk binds to the receptor assemblies through interactions of its pair of SH2 domains with ITAM motives of the receptor, which have been phosphorylated by Src-family kinases. These kinases also help to activate Syk by phosphorylation of its activation loop.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Syk
Applications Flow Cytometry
Classification Monoclonal
Clone 4D10.1
Concentration 5 μL/Test
Conjugate APC
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene Syk
Gene Accession No. P43405
Gene Alias DKFZp313N1010; EC 2.7.10.2; FLJ25043; FLJ37489; HGNC:11205; kinase Syk; KSYK; p72Syk; p72-Syk; ptk72; SP1; spleen associated tyrosine kinase; spleen tyrosine kinase; SYK; syk signaling; Sykb; tyrosine-kinase syk; tyrosine-protein kinase SYK
Gene Symbols Syk
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6850
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.