Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Symmetric Dimethyl Arginine Motif Rabbit anti-Human, Mouse, Rat, Clone: 8I0J1, Novus Biologicals™
SDP

Catalog No. NB167252 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167252 20 μg
NB167251 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB167252 Supplier Novus Biologicals Supplier No. NBP31607820UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Symmetric Dimethyl Arginine Motif Monoclonal antibody specifically detects Symmetric Dimethyl Arginine Motif in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Symmetric Dimethyl Arginine Motif
Applications Western Blot
Classification Monoclonal
Clone 8I0J1
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:1000
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias SDMA, Symmetric Dimethylarginine
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence containing methylated R.
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.