Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synapsin I Antibody, Novus Biologicals™
SDP

Catalog No. NB300104100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300104100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300104100 Supplier Novus Biologicals Supplier No. NB300104100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 42 publications

Synapsin I Polyclonal specifically detects Synapsin I in Human, Mouse, Rat, Guinea Pig, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating.

Specifications

Antigen Synapsin I
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:1000, Immunoprecipitation 1 ug per 200ug lysate, Immunohistochemistry-Frozen 1:10-1:500, Immunohistochemistry Free-Floating
Formulation 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative
Gene Accession No. P17599, P17600
Gene Alias SYN1a, SYN1b, Synapsin 1, Synapsin I, synapsin IBrain protein 4.1, synapsin-1, synaptic marker, synaptic markers immunohistochemistry, SYNI
Gene Symbols SYN1
Host Species Rabbit
Immunogen Native protein purified from bovine brain
Molecular Weight of Antigen 78 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Circadian Rhythm, Cognition and Behavior, Neuronal Cell Markers, Neuroscience, Neurotransmission, Pain Research, Sensory Systems, Vision
Primary or Secondary Primary
Gene ID (Entrez) 6853
Test Specificity Specific for the ∼78k synapsin I doublet in Western blots of rat brain extracts. Immunolabeling blocked by preadsorption of antibody with the protein used to generate the antibody.
Target Species Human, Mouse, Rat, Bovine, Guinea Pig, Primate
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.