Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synapsin I, p Ser9 Antibody, Novus Biologicals™
SDP

Catalog No. NB300180 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300180 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300180 Supplier Novus Biologicals Supplier No. NB300180
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Synapsin I Polyclonal specifically detects Synapsin I in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Synapsin I (p Ser9)
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:500
Formulation 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative
Gene Accession No. P09951
Gene Alias phospho Synapsin, SYN1a, SYN1b, Synapsin 1, synapsin IBrain protein 4.1, synapsin-1, SYNI
Gene Symbols SYN1
Host Species Rabbit
Immunogen Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser9 conjugated to KLH. Accession # P17599
Molecular Weight of Antigen 78 kDa
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Neuronal Cell Markers, Neuroscience, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 6853
Test Specificity Specific for ∼78k synapsin I doublet protein phosphorylated at Ser9. The antibody also weakly labels the ∼55k synapsin II protein which has a similar phosphorylation site to that of Ser9 on synapsin I. Immunolabeling is blocked by preadsorption of the antibody with the phosphopeptide used as antigen but not by the corresponding dephosphopeptide. Immunolabeling is also completely eliminated by treatment with lambda phosphatase.
Target Species Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.