Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synapsin II Antibody, Novus Biologicals™
SDP

Catalog No. NB018067 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018067 0.025 mg
NB12013258 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018067 Supplier Novus Biologicals Supplier No. NB120132580.025MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Synapsin II Polyclonal specifically detects Synapsin II in Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Synapsin II
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunoprecipitation 1:100
Formulation PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide
Gene Alias synapsin IISYNII, synapsin-2, SYNIIa, SYNIIb
Gene Symbols SYN2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a portion of rat Synapsin II. The sequence is completely conserved in mouse.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6854
Test Specificity This detects ∼74 and ∼54 kDa proteins, corresponding to the apparent molecular masses of synapsin IIa and IIb on SDS-PAGE immunoblots, in samples from mouse and rat origins.
Target Species Rat, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.