Learn More
Description
Specifications
Specifications
| Antigen | Synapsin II |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000, Immunoprecipitation 1:100 |
| Formulation | PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide |
| Gene Alias | synapsin IISYNII, synapsin-2, SYNIIa, SYNIIb |
| Gene Symbols | SYN2 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide corresponding to a portion of rat Synapsin II. The sequence is completely conserved in mouse. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
