Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synapsin I (p Ser9) Rabbit anti-Human, Mouse, Rat, Clone: SN63-04, Novus Biologicals™
SDP

Catalog No. NBP267662 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267662 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267662 Supplier Novus Biologicals Supplier No. NBP267662

Rabbit Monoclonal Antibody

Synapsin I Monoclonal specifically detects Synapsin I in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Synapsin I (p Ser9)
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SN63-04
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias SYN1a, SYN1b, Synapsin 1, synapsin IBrain protein 4.1, synapsin-1, SYNI
Gene Symbols SYN1
Host Species Rabbit
Immunogen Synthetic phospho-peptide corresponding to residues surrounding Ser9 of Human Synapsin Iaa 1-50/ 705. (SwissProt: P17600 Human; SwissProt: O88935 Mouse; SwissProt: P09951 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Cellular Markers, Circadian Rhythm, Cognition and Behavior, Neuronal Cell Markers, Neuroscience, Neurotransmission, Pain Research, Phospho Specific, Sensory Systems, Vision
Primary or Secondary Primary
Gene ID (Entrez) 6853
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.