Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synaptophysin Antibody (7H12), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP147483C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP147483C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP147483C Supplier Novus Biologicals Supplier No. NBP147483C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Synaptophysin Monoclonal specifically detects Synaptophysin in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Synaptophysin
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 7H12
Conjugate DyLight 650
Dilution Western Blot, ELISA, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Major synaptic vesicle protein p38, MRX, MRXSYP, synaptophysin
Gene Symbols SYP
Host Species Mouse
Immunogen Purified recombinant fragment of human Synaptophysin expressed in E. coli. [UniProt# P08247]
Purification Method Ammonium sulfate precipitation
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Membrane Vesicle Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6855
Target Species Human, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.