Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synaptophysin Antibody (SP11), Novus Biologicals™
SDP

Catalog No. NB12016659 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB12016659 0.5 mL
NB018076 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB12016659 Supplier Novus Biologicals Supplier No. NB12016659
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody has been used in 5 publications

Synaptophysin Monoclonal specifically detects Synaptophysin in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Synaptophysin
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SP11
Conjugate Unconjugated
Dilution Immunohistochemistry 1:25-1:50, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:25-1:50
Formulation Tissue culture supernatant with 0.05% Sodium Azide
Gene Alias Major synaptic vesicle protein p38, MRX, MRXSYP, synaptophysin
Gene Symbols SYP
Host Species Rabbit
Immunogen Synthetic peptide of human Synaptophysin.
Molecular Weight of Antigen 38 kDa
Purification Method Tissue culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Membrane Vesicle Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6855
Test Specificity This antibody recognizes a protein of 38kDa, identified as synaptophysin. It labels normal neuroendocrine cells of human adrenal medulla, carotid body, skin, pituitary gland, thyroid, lung, pancreas, gastrointestinal mucosa, Paneth's cells in the gastrointestinal tract and gastric parietal cells. Neurons in the brain, spinal cord, and retina are also labeled. In combination with anti-chromogranin A and anti-NSE, anti-synaptophysin is very useful in the identification of normal neuroendocrine cells and neuroendocrine neoplasms.
Target Species Human, Mouse
Content And Storage Store at 4°C.
Form Supernatant
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.