Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synaptophysin Antibody (SP11), Novus Biologicals™

Rabbit Monoclonal Antibody has been used in 5 publications
$304.50 - $646.00
Specifications
Antigen | Synaptophysin |
---|---|
Clone | SP11 |
Applications | Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Immunohistochemistry |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018076
![]() |
Novus Biologicals
NB120166590.1ML |
0.1 mL |
Each for $304.50
|
|
|||||
NB12016659
![]() |
Novus Biologicals
NB12016659 |
0.5 mL |
Each for $646.00
|
|
|||||
Description
Synaptophysin Monoclonal specifically detects Synaptophysin in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Synaptophysin | |
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Membrane Vesicle Markers, Stem Cell Markers | |
Major synaptic vesicle protein p38, MRX, MRXSYP, synaptophysin | |
SYP | |
IgG | |
Tissue culture supernatant | |
38 kDa |
SP11 | |
Monoclonal | |
Supernatant | |
RUO | |
Mouse, Human | |
6855 | |
Synthetic peptide of human Synaptophysin. | |
Primary | |
Store at 4°C. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title