Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synaptophysin Antibody (SP15), Novus Biologicals™
SDP

Catalog No. NBP19745905 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
0.2 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP19745905 0.05 mL
NB018770 0.025 mL
NBP19745902 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP19745905 Supplier Novus Biologicals Supplier No. NBP1974590.05ML

Mouse Monoclonal Antibody has been used in 3 publications

Synaptophysin Monoclonal specifically detects Synaptophysin in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Synaptophysin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SP15
Conjugate Unconjugated
Dilution Western Blot 1:10000, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide
Gene Alias Major synaptic vesicle protein p38, MRX, MRXSYP, synaptophysin
Gene Symbols SYP
Host Species Mouse
Immunogen Synaptic vesicle-containing fractions immunoprecipitated from human brain homogenates using anti-human Synaptophysin mAb antibodies.
Purification Method Ammonium sulfate precipitation
Quantity 0.05 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6855
Target Species Human, Mouse, Rat, Bovine
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.