Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synaptophysin Antibody (SP15), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 3 publications
$280.00 - $402.00
Specifications
Antigen | Synaptophysin |
---|---|
Clone | SP15 |
Dilution | Western Blot 1:10000, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin 1:10-1:500 |
Applications | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018770
![]() |
Novus Biologicals
NBP1974590.025ML |
0.025 mL |
Each for $280.00
|
|
|||||
NBP19745905
![]() |
Novus Biologicals
NBP1974590.05ML |
0.05 mL |
Each for $402.00
|
|
|||||
NBP19745902
![]() |
Novus Biologicals
NBP1974590.2ML |
0.2 mL | N/A | N/A | N/A | ||||
Description
Synaptophysin Monoclonal specifically detects Synaptophysin in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Synaptophysin | |
Western Blot 1:10000, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
6855 | |
Synaptic vesicle-containing fractions immunoprecipitated from human brain homogenates using anti-human Synaptophysin mAb antibodies. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
SP15 | |
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Mouse, Rat, Human, Bovine | |
Major synaptic vesicle protein p38, MRX, MRXSYP, synaptophysin | |
SYP | |
IgM | |
Ammonium sulfate precipitation |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title