Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synaptophysin Antibody (SYP/4503R), Biotin, Novus Biologicals™
SDP

Catalog No. NB382773 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB382773 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB382773 Supplier Novus Biologicals Supplier No. NBP308213B
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Synaptophysin Monoclonal antibody specifically detects Synaptophysin in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen Synaptophysin
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SYP/4503R
Conjugate Biotin
Formulation PBS
Gene Alias Major synaptic vesicle protein p38, MRX, MRXSYP, synaptophysin
Host Species Rabbit
Immunogen Recombinant fragment (around aa 224-313) of human Synaptophysin (SYP) protein (exact sequence is proprietary) (Uniprot: IDP08247)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Membrane Vesicle Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6855
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.