Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synaptotagmin 1 Antibody (HL1654) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB294911 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB294911 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB294911 Supplier Novus Biologicals Supplier No. NBP325706
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Synaptotagmin 1 Monoclonal antibody specifically detects Synaptotagmin 1 in Human, Mouse, Rat, Drosophila samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Synaptotagmin 1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone HL1654
Conjugate Unconjugated
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias DKFZp781D2042, p65, SVP65, synaptotagmin Isynaptotagmin 1, synaptotagmin-1, SYT, SytI
Host Species Rabbit
Immunogen Recombinant fragment of human Synaptotagmin 1.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Membrane Vesicle Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6857
Target Species Human, Mouse, Rat, Drosophila
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.