Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synaptotagmin 1, p Thr202 Antibody, Novus Biologicals™
SDP

Catalog No. NB300235 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300235 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300235 Supplier Novus Biologicals Supplier No. NB300235
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Synaptotagmin 1 Polyclonal specifically detects Synaptotagmin 1 in Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Synaptotagmin 1 (p Thr202)
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:400
Formulation 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative
Gene Accession No. P21707
Gene Alias DKFZp781D2042, p65, SVP65, synaptotagmin Isynaptotagmin 1, synaptotagmin-1, SYT, SytI
Gene Symbols SYT1
Host Species Rabbit
Immunogen Synthetic phospho-peptide corresponding to amino acid residues surrounding Thr202 conjugated to KLH. Accession # P21707
Molecular Weight of Antigen 60 kDa
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Membrane Vesicle Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 6857
Test Specificity Specific for the ∼60k - 62k synaptotagmin protein phosphorylated at Thr202. The immunolabeling is completely eliminated by lambda-phosphatase treatment. In some lysates and/or various tissues, additional bands may be seen at ∼ 45k, 75k and 150k.
Target Species Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.