Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synaptotagmin 4 Antibody (OTI4H3), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP274415FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274415FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274415FR Supplier Novus Biologicals Supplier No. NBP274415FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Synaptotagmin 4 Monoclonal specifically detects Synaptotagmin 4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Synaptotagmin 4
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI4H3
Conjugate DyLight 680
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias HsT1192, KIAA1342sytIV, synaptotagmin 4, synaptotagmin IVsynaptotagmin-4, SytIV
Gene Symbols SYT4
Host Species Mouse
Immunogen Full length recombinant protein of human Synaptotagmin 4 (NP_065834) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Membrane Vesicle Markers
Primary or Secondary Primary
Gene ID (Entrez) 6860.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.