Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SYNC Antibody (OTI1E7), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. N274416A647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274416A647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274416A647 Supplier Novus Biologicals Supplier No. NBP274416AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SYNC Monoclonal specifically detects SYNC in Human samples. It is validated for Western Blot.

Specifications

Antigen SYNC
Applications Western Blot
Classification Monoclonal
Clone OTI1E7
Conjugate Alexa Fluor 647
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias SYNC syncoilin, intermediate filament protein, SYNC1, SYNCOILIN
Gene Symbols SYNC
Host Species Mouse
Immunogen Full length recombinant protein of human SYNC (NP_110413) produced in SF9 cell.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 81493.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.