Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syndecan-1/CD138 Antibody (359103R), FITC, Novus Biologicals™
SDP

Catalog No. NB364169 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB364169 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB364169 Supplier Novus Biologicals Supplier No. FAB2780RF
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Syndecan-1/CD138 Monoclonal antibody specifically detects Syndecan-1/CD138 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen Syndecan-1/CD138
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 359103R
Conjugate FITC
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias CD138, CD138 antigen, SDC, SYND1heparan sulfate proteoglycan fibroblast growth factor receptor, syndecan, syndecan 1, syndecan proteoglycan 1, syndecan-1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Syndecan-1/CD138, Gln18-Glu251, Accession # NP_002988
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers
Primary or Secondary Primary
Gene ID (Entrez) 6382
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.