Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syndecan-1/CD138 Antibody (B-A38), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB388667 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB388667 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB388667 Supplier Novus Biologicals Supplier No. NBP314568AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Syndecan-1/CD138 Monoclonal antibody specifically detects Syndecan-1/CD138 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry

Specifications

Antigen Syndecan-1/CD138
Applications Flow Cytometry, ELISA, Immunohistochemistry
Classification Monoclonal
Clone B-A38
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias CD138, CD138 antigen, SDC, SYND1heparan sulfate proteoglycan fibroblast growth factor receptor, syndecan, syndecan 1, syndecan proteoglycan 1, syndecan-1
Host Species Mouse
Immunogen U266 cell line
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers
Primary or Secondary Primary
Gene ID (Entrez) 6382
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.