Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syndecan-2/CD362 Antibody (305515), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB363543 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB363543 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB363543 Supplier Novus Biologicals Supplier No. FAB2965K
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Syndecan-2/CD362 Monoclonal antibody specifically detects Syndecan-2/CD362 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen Syndecan-2/CD362
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 305515
Conjugate DyLight 488
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD362 antigen, fibroglycan, heparan sulfate proteoglycan 1, cell surface-associated, Heparan sulfate proteoglycan core protein, HSPG, HSPG1syndecan proteoglycan 2, SYND2cell surface-associated heparan sulfate proteoglycan 1, syndecan 2, syndecan-2
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Syndecan-2/CD362, Glu19-Gly144, Accession # AAH49836.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 6383
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.