Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syndecan-4 Antibody (008), HRP, Novus Biologicals™
SDP

Catalog No. NB076672 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB076672 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB076672 Supplier Novus Biologicals Supplier No. NBP290699H
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Syndecan-4 Monoclonal specifically detects Syndecan-4 in Mouse samples. It is validated for ELISA.

Specifications

Antigen Syndecan-4
Applications ELISA
Classification Monoclonal
Clone 8
Conjugate HRP
Formulation PBS
Gene Alias amphiglycan, MGC22217, ryudocan, Ryudocan core protein, SYND4ryudocan amphiglycan, syndecan 4, syndecan 4 (amphiglycan, ryudocan), syndecan proteoglycan 4, syndecan-4
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Syndecan-4 (O35988; Met1-Val146).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Signaling, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6385
Test Specificity No cross-reactivity in ELISA with: Mouse SDC1
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.