Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syndecan-4 Antibody (336304), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB348642 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB348642 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB348642 Supplier Novus Biologicals Supplier No. FAB29181MFV500
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Syndecan-4 Monoclonal specifically detects Syndecan-4 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen Syndecan-4
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 336304
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias amphiglycan, MGC22217, ryudocan, Ryudocan core protein, SYND4ryudocan amphiglycan, syndecan 4, syndecan 4 (amphiglycan, ryudocan), syndecan proteoglycan 4, syndecan-4
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Syndecan-4, Ser20-Glu145, Accession # P31431
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Signaling, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6385
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.