Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syndecan-4 Antibody, Novus Biologicals™
SDP

Catalog No. NB017971 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017971 0.025 mg
NB11041551 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017971 Supplier Novus Biologicals Supplier No. NB110415510.025MG

Rabbit Polyclonal Antibody

Syndecan-4 Polyclonal specifically detects Syndecan-4 in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Syndecan-4
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS (pH 7.2), 0.5% BSA, 30% Glycerol with 0.01% Thimerosal
Gene Accession No. P31431
Gene Alias amphiglycan, MGC22217, ryudocan, Ryudocan core protein, SYND4ryudocan amphiglycan, syndecan 4, syndecan 4 (amphiglycan, ryudocan), syndecan proteoglycan 4, syndecan-4
Gene Symbols SDC4
Host Species Rabbit
Immunogen Synthetic peptide surrounding amino acid 184 of human Syndecan-4.
Molecular Weight of Antigen 22 kDa
Purification Method Affinity purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6385
Target Species Chicken, Mouse, Rat, Human
Content And Storage Store at -20 degreesC.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.