Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SYNE1 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB052277 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB052277 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB052277 Supplier Novus Biologicals Supplier No. NBP298081G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SYNE1 Polyclonal specifically detects SYNE1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SYNE1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ARCA1, C6orf98, CPG2, dJ45H2.2, EC 4.2.1.49, EC 4.3.1.3, EDMD4, Enaptin, KIAA0796chromosome 6 open reading frame 98, KIAA1262FLJ41140, KIAA17568B, Myne-1, MYNE1DKFZp781J13156, Myocyte nuclear envelope protein 1, nesprin 1, nesprin-1, Nuclear envelope spectrin repeat protein 1, SCAR8FLJ30878, spectrin repeat containing, nuclear envelope 1, Synaptic nuclear envelope protein 1, synaptic nuclei expressed gene 1, Syne-1, SYNE-1B
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SYNE1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Developmental Biology
Primary or Secondary Primary
Gene ID (Entrez) 23345
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.