Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent BioTek Synergy H1 Hybrid Multi-Mode Reader PROMO

Catalog No. BTH1MGPM Shop All Agilent Technologies Inc. Products
Click to view available options
Model:
H1M
H1MG
This item is not returnable. View return policy
This item is not returnable. View return policy

BioTek Synergy H1 is a modular multi-mode microplate reader, with monochromator-based optics and filter-based optics. For Research Use Only. Not for use in diagnostic procedures.

BioTek Synergy H1 is a modular multi-mode microplate reader, with monochromator-based optics and filter-based optics. The patented Hybrid Technology offers flexibility and sensitivity across a broad range of applications. The modular platform allows upgrading to expand functionality as your laboratory workflows change

Synergy H1 offers continuously variable bandwidth monochromators for fluorescence excitation and emission wavelength selection. Fluorescence bandwidth can be set between 9 and 50 nm, in 1 nm increments, allowing users to fully optimize reader settings to drive the best assay performance compared to fixed bandwidth systems.

TRUSTED_SUSTAINABILITY

Specifications

Absorbance Range Absorbance: 230 to 999 nm (1 nm increments)
Accuracy Absorbance: ±2 nm; Dispenser: ±1 μL or 2%
Certifications/Compliance CE and TUV marked. RoHS compliant.
CO2 Concentration Range 0 to 20%
Depth (Metric) 46.4 cm
Compatibility BioStack and Third Party Automation Compatible
Depth (English) 18.3 in.
Incubator Temperature 4-Zone Incubation to 45°C
Measurement Speed 11 sec. (96 wells), 22 sec. (384 wells)
Shaking Linear, Orbital, Double Orbital
Dynamic Range 0 to 4 OD
Dispense Volume 5 to 1000 μL, 1 μL increments
Height (English) 13 in.
Frequency 50/60 Hz
Height (Metric) 33 cm
Includes Compatible with gas controler; Monochromator fluorescence; Monochromator absorbance; Full-light luminescence; Time resolved fluorescence
Measurement Technology Proprietary Hybrid Technology
Model H1MG
No. of Dispensers 2 Syringe Pumps
Product Type Multi-Mode Microplate Reader
Power Consumption 130 W max.
Resolution ±0.1% (CO2 and O2)
Sensitivity 2.5 to 4 pM monochromators, 0.25 pM filters fluorescence, 20 amol ATP monochromator, 10 amol ATP filter luminescence, 1.2 mP fluorescence polarization, Europium 40 fM with filters, 1200 fM with monos time resolved fluorescence
Wavelength Range Fluorescence Intensity Monochromator: 250 to 700 nm (900 nm option), Fluorescence Polarization: 280 to 700 nm (850 nm option)
Weight (English) <55 lb.
Weight (Metric) 25 kg
Width (English) 14.8 in.
Width (Metric) 37.5 cm
Plate Type 6- to 384-well Plates
Detector Type Photodiode
Software Type Gen5 Microplate Reader and Imager Software
Light Source Xenon Flash
Reading Options End Point
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.