Learn More
Agilent BioTek Synergy H1 Hybrid Multi-Mode Reader PROMO
Description
BioTek Synergy H1 is a modular multi-mode microplate reader, with monochromator-based optics and filter-based optics. The patented Hybrid Technology offers flexibility and sensitivity across a broad range of applications. The modular platform allows upgrading to expand functionality as your laboratory workflows change
Synergy H1 offers continuously variable bandwidth monochromators for fluorescence excitation and emission wavelength selection. Fluorescence bandwidth can be set between 9 and 50 nm, in 1 nm increments, allowing users to fully optimize reader settings to drive the best assay performance compared to fixed bandwidth systems.

Specifications
Specifications
Absorbance Range | 230 to 999 nm (1 nm increments) |
Accuracy | Absorbance: ±2 nm; Dispenser: ±1 μL or 2% |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
CO2 Concentration Range | 0 to 20% |
Depth (Metric) | 46.4 cm |
Compatibility | BioStack and Third Party Automation Compatible |
Depth (English) | 18.3 in. |
Incubator Temperature | 4-Zone Incubation to 45°C |
Measurement Speed | 11 sec. (96 wells), 22 sec. (384 wells) |
Shaking | Linear, Orbital, Double Orbital |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.