Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent BioTek Synergy LX Multi-Mode Reader, IVD
GREENER_CHOICE

Catalog No. p-7236170 Shop All Agilent Technologies Inc. Products
Click to view available options
Model:
SLXA
SLXATS
SLXF
SLXFA
SLXFATS
SLXFTS
Includes:
Absorbance
Absorbance, Fluorescence, Luminescence
Absorbance, Fluorescence, Luminescence, Touchscreen
Absorbance, Luminescence, Touchscreen
Fluorescence, Luminescence, Touchscreen
Fluorescence; Luminescence
This item is not returnable. View return policy

Catalog No. BTSLXASI

Supplier: Agilent BioTek SLXASI

Only null left
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy

The BioTek Synergy LX multi-mode reader automates many common microplate assays. For In Vitro Diagnostic Use.

The BioTek Synergy LX multi-mode reader automates many common microplate assays. The high-quality optic design ensures excellent data in absorbance, fluorescence, and luminescence detection modes. This affordable system is ideal for many common assays including nucleic acid and protein quantification, ELISA, BCA, Bradford, and cell viability

The data display, output to a USB flash drive, printer, or Gen5 software make Synergy LX a versatile assay workstation. The large touch screen user interface and onboard software simplify programming and save time in operation of microplate and microvolume assays.

TRUSTED_SUSTAINABILITY

Specifications

Absorbance Range 200 to 900 nm
Accuracy Wavelength: ±2 nm; Optical Density: <1% at 2.0 OD, <3% at 2.5 OD
Certifications/Compliance CE and TUV marked. RoHS compliant.
Bandwidth ≤5 nm
Depth (Metric) 38.1 cm
Depth (English) 15 in.
Measurement Speed 24 sec. (96 wells), 76 sec. (384 wells)
Shaking Linear, Orbital, Double Orbital
Filters Monochromator Bandpass
For Use With (Application) Nucleic acid and protein quantification, ELISA, BCA, Bradford and cell viability
Height (English) 12 in.
Frequency 50/60 Hz
Height (Metric) 30.5 cm
Linearity <1% from 0 to 2.5 OD
Includes Absorbance
Model SLXA
Product Type Multi-Mode Microplate Reader
Power Consumption 24 VDC power supply compatible with 100/240 VAC at 50/60 Hz, 60 W (max.)
Resolution 0.001 OD
Sensitivity Fluorescence Intensity: Fluorescein 2 pM; Luminesence: 10 amol ATP
USB Connections 2
Software Language Versions English
Warranty 1 year
Voltage 100/240 V
Weight (English) 27 lb.
Weight (Metric) 12.3 kg
Width (English) 15 in.
Width (Metric) 38.1 cm
Detector Type Photodiode, PMT
Software Type Full data analysis and reporting (under Gen5 control)
Light Source Xenon Flash
Reading Options End Point, Kinetic, Area Scanning, Absorbance Spectral Scanning
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.