Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SynGAP Antibody, Novus Biologicals™
SDP

Catalog No. NBP227541 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227541 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP227541 Supplier Novus Biologicals Supplier No. NBP227541
Add to Cart
Add to Cart

Goat Polyclonal Antibody

SynGAP Polyclonal specifically detects SynGAP in Mouse samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen SynGAP
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.03 - 0.1 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_006763.2
Gene Alias DKFZp761G1421, KIAA1938135kDa, MRD5, synaptic Ras GTPase activating protein 1, synaptic Ras GTPase activating protein 1 homolog (rat), Synaptic Ras-GAP 1
Gene Symbols SYNGAP1
Host Species Goat
Immunogen Peptide with sequence C-ESAHIEREEYKLKEY corresponding to internal region (near C-Terminus) according to NP_006763.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cognition and Behavior, Neuroscience, Vision
Primary or Secondary Primary
Gene ID (Entrez) 8831
Test Specificity Expected from sequence similarity: Human, Rat, Pig, Dog, Cow.
Target Species Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.