Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SynGAP Rabbit anti-Human, Mouse, Rat, Clone: 8M2S9, Novus Biologicals™
SDP

Catalog No. NB166670 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166670 20 μg
NB166669 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB166670 Supplier Novus Biologicals Supplier No. NBP31578720UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SynGAP Monoclonal antibody specifically detects SynGAP in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen SynGAP
Applications Western Blot
Classification Monoclonal
Clone 8M2S9
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias DKFZp761G1421, KIAA1938135kDa, MRD5, synaptic Ras GTPase activating protein 1, synaptic Ras GTPase activating protein 1 homolog (rat), Synaptic Ras-GAP 1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1244-1343 of human SynGAP (Q96PV0).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cognition and Behavior, Neuroscience, Vision
Primary or Secondary Primary
Gene ID (Entrez) 8831
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.